This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05063427_m1
See other CNBP GE Assays ›
Gene Symbol:
Cnbp
Gene Name:
CCHC-type zinc finger nucleic acid binding protein
Gene Aliases:
GW7_06224
Chromosome Location:
Chr.Un|NW_004624943: 943412 - 956011 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05063427_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cnbp
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001279877.1 NP_001266806.1 4 - 5 583 70
XM_013069290.2 XP_012924744.2 4 - 5 431 70
XM_013069293.2 XP_012924747.1 4 - 5 631 70
XM_021252195.1 XP_021107854.1 3 - 4 433 70
XM_021252196.1 XP_021107855.1 3 - 4 415 70
XM_021252197.1 XP_021107856.1 3 - 4 412 70
XM_021252198.1 XP_021107857.1 3 - 4 406 70
XM_021252199.1 XP_021107858.1 3 - 4 403 70
XM_021252200.1 XP_021107859.1 4 - 5 611 70
XM_021252201.1 XP_021107860.1 4 - 5 602 70
GenBank mRNA HQ326654.1 - 4 - 5 583 70

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

nucleic acid metabolism protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0