This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05065076_g1
See other SLC7A7 GE Assays ›
Gene Symbol:
Slc7a7
Gene Name:
solute carrier family 7 member 7
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_004624820: 9708481 - 9757251 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05065076_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc7a7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004864205.2 XP_004864262.1 6 - 7 1217 58
XM_004864206.2 XP_004864263.1 6 - 7 1224 58
XM_021243903.1 XP_021099562.1 6 - 7 1240 58
XM_021243904.1 XP_021099563.1 8 - 9 1396 58
XM_021243905.1 XP_021099564.1 6 - 7 1063 58
XM_021243906.1 XP_021099565.1 6 - 7 1240 58

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

transporter

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0