This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05069552_m1
See other PNPLA4 GE Assays ›
Gene Symbol:
Pnpla4
Gene Name:
patatin like phospholipase domain containing 4
Gene Aliases:
GW7_12685
Chromosome Location:
Chr.Un|NW_004624834: 5497109 - 5530037 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05069552_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pnpla4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004867352.3 XP_004867409.1 6 - 7 739 64
XM_004867353.3 XP_004867410.1 6 - 7 843 64
XM_004867354.3 XP_004867411.1 5 - 6 671 64
XM_004867355.2 XP_004867412.1 6 - 7 726 64
XM_021246410.1 XP_021102069.1 6 - 7 737 64
XM_021246411.1 XP_021102070.1 5 - 6 670 64

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

hydrolase lipase phospholipase metabolite interconversion enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0