This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05070192_m1
See other LOC101720124 GE Assays ›
Gene Symbol:
LOC101720124
Gene Name:
aldose reductase
Gene Aliases:
Akr1b1, GW7_05923
Chromosome Location:
Chr.Un|NW_004624783: 1945203 - 1958073 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05070192_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC101720124
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004856310.3 XP_004856367.1 4 - 5 519 67
XM_021237598.1 XP_021093257.1 4 - 5 348 67

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Probe spans exons

Panther Classification:

Molecular Function -

oxidoreductase reductase metabolite interconversion enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-55658555d6-pnd5g:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline