This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05073156_g1
See other EIF4E2 GE Assays ›
Gene Symbol:
Eif4e2
Gene Name:
eukaryotic translation initiation factor 4E family member 2
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_004624843: 5303178 - 5332228 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05073156_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Eif4e2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004868068.3 XP_004868125.1 3 - 4 453 65
XM_004868069.3 XP_004868126.1 3 - 4 404 65
XM_004868070.3 XP_004868127.1 3 - 4 453 65
XM_004868071.3 XP_004868128.1 3 - 4 453 65
XM_004868072.3 XP_004868129.1 3 - 4 400 65
XM_004868073.3 XP_004868130.1 3 - 4 353 65

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

translation factor translation initiation factor translational protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0