This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05073619_g1
See other CXXC5 GE Assays ›
Gene Symbol:
Cxxc5
Gene Name:
CXXC finger protein 5
Gene Aliases:
GW7_08869
Chromosome Location:
Chr.Un|NW_004624743: 32450624 - 32485249 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05073619_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cxxc5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004841845.2 XP_004841902.1 3 - 4 1364 86
XM_004841847.3 XP_004841904.1 3 - 4 1260 86
XM_004841848.3 XP_004841905.1 2 - 3 1159 86
XM_004841849.3 XP_004841906.1 3 - 4 1264 86
XM_004841850.3 XP_004841907.1 2 - 3 1163 86
XM_021255912.1 XP_021111571.1 3 - 4 1312 86

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rr9bx:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0