This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05078370_mH
See other ELAVL2 GE Assays ›
Gene Symbol:
Elavl2
Gene Name:
ELAV like RNA binding protein 2
Gene Aliases:
GW7_06083
Chromosome Location:
Chr.Un|NW_004624736: 27692281 - 27857560 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05078370_mH
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Elavl2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004838359.3 XP_004838416.1 3 - 4 624 83
XM_004838360.3 XP_004838417.1 3 - 4 496 83
XM_013069755.2 XP_012925209.2 3 - 4 432 83
XM_013069757.2 XP_012925211.1 4 - 5 545 83
XM_013069761.2 XP_012925215.1 3 - 4 854 83
XM_021252910.1 XP_021108569.1 4 - 5 586 83
XM_021252911.1 XP_021108570.1 3 - 4 429 83
XM_021252912.1 XP_021108571.1 3 - 4 432 83
XM_021252913.1 XP_021108572.1 4 - 5 974 83
XM_021252914.1 XP_021108573.1 5 - 6 725 83
XM_021252915.1 XP_021108574.1 4 - 5 635 83
XM_021252916.1 XP_021108575.1 4 - 5 561 83
XM_021252917.1 XP_021108576.1 4 - 5 477 83
XM_021252918.1 XP_021108577.1 3 - 4 432 83
XM_021252919.1 XP_021108578.1 5 - 6 788 83
XM_021252920.1 XP_021108579.1 4 - 5 949 83
XM_021252921.1 XP_021108580.1 4 - 5 610 83
XM_021252922.1 XP_021108581.1 4 - 5 453 83
XM_021252923.1 XP_021108582.1 5 - 6 786 83
XM_021252924.1 XP_021108583.1 4 - 5 947 83
XM_021252925.1 XP_021108584.1 4 - 5 717 83
XM_021252926.1 XP_021108585.1 3 - 4 518 83
XM_021252927.1 XP_021108586.1 4 - 5 696 83
XM_021252928.1 XP_021108587.1 3 - 4 857 83
XM_021252929.1 XP_021108588.1 3 - 4 565 83
XM_021252930.1 XP_021108589.1 3 - 4 457 83
XM_021252931.1 XP_021108590.1 3 - 4 360 83

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

translation initiation factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7gdsv:80/100.66.79.88:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0