This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05081053_g1
See other HYI GE Assays ›
Gene Symbol:
Hyi
Gene Name:
hydroxypyruvate isomerase (putative)
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_004624892: 2753167 - 2755739 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05081053_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Hyi
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004871868.3 XP_004871925.2 1 - 2 622 63
XM_004871870.3 XP_004871927.1 1 - 2 253 63
XM_004871873.3 XP_004871930.1 1 - 2 251 63
XM_004871875.3 XP_004871932.2 1 - 2 622 63
XM_013068167.2 XP_012923621.1 1 - 2 258 63
XM_021249907.1 XP_021105566.1 1 - 2 265 63
XM_021249908.1 XP_021105567.1 1 - 2 621 63
XM_021249909.1 XP_021105568.1 1 - 2 252 63
XM_021249911.1 XP_021105570.1 1 - 2 621 63

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

isomerase metabolite interconversion enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline