This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05081537_g1
See other DUSP15 GE Assays ›
Gene Symbol:
Dusp15
Gene Name:
dual specificity phosphatase 15
Gene Aliases:
GW7_07708
Chromosome Location:
Chr.Un|NW_004624741: 651550 - 660876 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05081537_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dusp15
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004840592.3 XP_004840649.1 2 - 3 178 64
XM_004840593.3 XP_004840650.1 2 - 3 178 64
XM_021254911.1 XP_021110570.1 2 - 3 178 64
XM_021254912.1 XP_021110571.1 2 - 3 156 64
XM_021254913.1 XP_021110572.1 2 - 3 148 64
XM_021254914.1 XP_021110573.1 2 - 3 141 64

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_021254914.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

protein phosphatase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0