This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05082834_g1
See other LIMA1 GE Assays ›
Gene Symbol:
Lima1
Gene Name:
LIM domain and actin binding 1
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_004624816: 2052602 - 2146473 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05082834_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Lima1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004862983.3 XP_004863040.1 4 - 5 771 66
XM_004862984.3 XP_004863041.1 5 - 6 890 66
XM_004862985.3 XP_004863042.1 4 - 5 771 66
XM_004862987.3 XP_004863044.1 4 - 5 771 66
XM_021243041.1 XP_021098700.1 5 - 6 837 66
XM_021243042.1 XP_021098701.1 1 - 2 607 66

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

actin or actin-binding cytoskeletal protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-w8bxd:80/100.66.79.51:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0