This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05085098_m1
See other SS18 GE Assays ›
Gene Symbol:
Ss18
Gene Name:
SS18, nBAF chromatin remodeling complex subunit
Gene Aliases:
GW7_07896
Chromosome Location:
Chr.Un|NW_004624770: 6256552 - 6343250 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05085098_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ss18
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004853140.3 XP_004853197.1 5 - 6 816 68
XM_013075276.2 XP_012930730.1 5 - 6 809 68
XM_021265134.1 XP_021120793.1 5 - 6 821 68
XM_021265135.1 XP_021120794.1 4 - 5 614 68
XM_021265136.1 XP_021120795.1 5 - 6 834 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

transcription cofactor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-w9t66:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline