Assay Details
| Gene Symbol: Atrip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004833942.2 | XP_004833999.1 | 12 - 13 | 2423 | 63 | |
| XM_021256865.1 | XP_021112524.1 | 12 - 13 | 2439 | 63 | ||
| XM_021256951.1 | XP_021112610.1 | 12 - 13 | 2095 | 63 | ||
More Information
Set Membership: |
|

Atrip
Genomic Map