This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05086524_m1
See other SLC7A10 GE Assays ›
Gene Symbol:
Slc7a10
Gene Name:
solute carrier family 7 member 10
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_004624794: 9876617 - 9892224 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05086524_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc7a10
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004858919.3 XP_004858976.1 7 - 8 1180 78
XM_021239568.1 XP_021095227.1 9 - 10 1382 78
XM_021239569.1 XP_021095228.1 9 - 10 1362 78
XM_021239570.1 XP_021095229.1 6 - 7 1092 78
XM_021239571.1 XP_021095230.1 6 - 7 1092 78
XM_021239572.1 XP_021095231.1 7 - 8 1183 78
XM_021239573.1 XP_021095232.1 8 - 9 1525 78
XM_021239574.1 XP_021095233.1 8 - 9 1768 78

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

transporter

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline