This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05086739_g1
See other YTHDF3 GE Assays ›
Gene Symbol:
Ythdf3
Gene Name:
YTH N6-methyladenosine RNA binding protein 3
Gene Aliases:
GW7_02254
Chromosome Location:
Chr.Un|NW_004624744: 28558988 - 28597212 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05086739_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ythdf3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004842127.3 XP_004842184.1 4 - 5 1980 86
XM_004842129.3 XP_004842186.1 5 - 6 2012 86
XM_004842130.3 XP_004842187.1 5 - 6 1931 86
XM_013071247.2 XP_012926701.1 5 - 6 2127 86
XM_013071249.2 XP_012926703.1 4 - 5 2012 86
XM_013071250.2 XP_012926704.1 4 - 5 1789 86

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0