This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05090326_g1
See other MPI GE Assays ›
Gene Symbol:
Mpi
Gene Name:
mannose phosphate isomerase
Gene Aliases:
GW7_01863
Chromosome Location:
Chr.Un|NW_004624894: 84531 - 93957 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05090326_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mpi
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004872032.3 XP_004872089.1 4 - 5 523 60
XM_021250046.1 XP_021105705.1 3 - 4 814 60
XM_021250047.1 XP_021105706.1 3 - 4 800 60
XM_021250048.1 XP_021105707.1 3 - 4 433 60
XM_021250049.1 XP_021105708.1 4 - 5 523 60
XM_021250050.1 XP_021105709.1 4 - 5 526 60
XM_021250051.1 XP_021105710.1 4 - 5 594 60
XM_021250052.1 XP_021105711.1 3 - 4 479 60
XM_021250053.1 XP_021105712.1 3 - 4 408 60
XM_021250054.1 XP_021105713.1 3 - 4 364 60
XM_021250055.1 XP_021105714.1 2 - 3 300 60
XM_021250056.1 XP_021105715.1 3 - 4 811 60

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

isomerase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-wt24k:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline