This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05093228_g1
See other NAA60 GE Assays ›
Gene Symbol:
Naa60
Gene Name:
N(alpha)-acetyltransferase 60, NatF catalytic subunit
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_004624824: 1255353 - 1289413 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05093228_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Naa60
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004864737.3 XP_004864794.1 3 - 4 639 102
XM_004864739.3 XP_004864796.1 3 - 4 428 102
XM_004864740.3 XP_004864797.1 3 - 4 402 102
XM_004864742.3 XP_004864799.1 3 - 4 652 102
XM_021244404.1 XP_021100063.1 4 - 5 504 102
XM_021244405.1 XP_021100064.1 3 - 4 270 102

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

acetyltransferase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0