This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05094350_m1
See other GLT8D2 GE Assays ›
Gene Symbol:
Glt8d2
Gene Name:
glycosyltransferase 8 domain containing 2
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_004624750: 4660521 - 4707258 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05094350_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Glt8d2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004844828.3 XP_004844885.1 2 - 3 107 72
XM_004844829.2 XP_004844886.1 2 - 3 234 72
XM_004844830.2 XP_004844887.1 3 - 4 340 72
XM_004844831.2 XP_004844888.1 2 - 3 224 72
XM_021258394.1 XP_021114053.1 2 - 3 223 72
XM_021258395.1 XP_021114054.1 2 - 3 99 72
XM_021258396.1 XP_021114055.1 2 - 3 73 72

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_004844829.2, XM_004844830.2, XM_004844831.2, XM_021258394.1, XM_021258395.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

glycosyltransferase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0