This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05097841_m1
See other FGF1 GE Assays ›
Gene Symbol:
Fgf1
Gene Name:
fibroblast growth factor 1
Gene Aliases:
GW7_10596
Chromosome Location:
Chr.Un|NW_004624774: 1778407 - 1867304 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05097841_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Fgf1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004854321.3 XP_004854378.1 3 - 4 706 68
XM_004854322.3 XP_004854379.1 3 - 4 713 68
XM_021266137.1 XP_021121796.1 3 - 4 716 68
XM_021266138.1 XP_021121797.1 4 - 5 607 68
XM_021266139.1 XP_021121798.1 4 - 5 978 68
XM_021266140.1 XP_021121799.1 3 - 4 584 68
XM_021266141.1 XP_021121800.1 2 - 3 986 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

growth factor intercellular signal molecule

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-4kcjf:80/100.66.78.36:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline