This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05098312_g1
See other PPP2R2B GE Assays ›
Gene Symbol:
Ppp2r2b
Gene Name:
protein phosphatase 2 regulatory subunit Bbeta
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_004624774: 6080890 - 6539197 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05098312_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ppp2r2b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004854350.3 XP_004854407.1 4 - 5 646 72
XM_004854351.3 XP_004854408.1 4 - 5 742 72
XM_004854352.3 XP_004854409.1 5 - 6 718 72
XM_004854353.3 XP_004854410.1 4 - 5 523 72
XM_004854354.3 XP_004854411.1 3 - 4 542 72
XM_004854355.3 XP_004854412.1 4 - 5 551 72
XM_004854356.3 XP_004854413.1 4 - 5 998 72
XM_013075737.2 XP_012931191.1 4 - 5 794 72
XM_021266166.1 XP_021121825.1 5 - 6 416 72
XM_021266167.1 XP_021121826.1 2 - 3 814 72

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

protein phosphatase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0