This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05098575_g1
See other TMEM25 GE Assays ›
Gene Symbol:
Tmem25
Gene Name:
transmembrane protein 25
Gene Aliases:
GW7_18664
Chromosome Location:
Chr.Un|NW_004624784: 13931563 - 13936664 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05098575_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tmem25
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004856703.3 XP_004856760.1 8 - 9 1526 92
XM_004856704.3 XP_004856761.1 8 - 9 1523 92
XM_004856705.3 XP_004856762.1 8 - 9 1452 92
XM_004856706.3 XP_004856763.1 7 - 8 1500 92
XM_004856707.3 XP_004856764.1 7 - 8 1391 92
XM_004856708.3 XP_004856765.1 8 - 9 1182 92
XM_004856709.3 XP_004856766.1 8 - 9 1209 92
XM_004856710.3 XP_004856767.1 8 - 9 1137 92
XM_021237691.1 XP_021093350.1 7 - 8 1320 92
XM_021237692.1 XP_021093351.1 6 - 7 1197 92
XM_021237693.1 XP_021093352.1 7 - 8 897 92

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0