This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05099404_g1
See other PCED1A GE Assays ›
Gene Symbol:
Pced1a
Gene Name:
PC-esterase domain containing 1A
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_004624741: 3953955 - 3960352 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05099404_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pced1a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004840685.3 XP_004840742.1 3 - 4 417 59
XM_004840689.3 XP_004840746.1 3 - 4 419 59
XM_021254978.1 XP_021110637.1 2 - 3 503 59
XM_021254979.1 XP_021110638.1 2 - 3 511 59
XM_021254980.1 XP_021110639.1 4 - 5 1206 59
XM_021254981.1 XP_021110640.1 3 - 4 1138 59

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-lcddp:80/100.66.76.150:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0