This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05100148_g1
See other DRAM2 GE Assays ›
Gene Symbol:
Dram2
Gene Name:
DNA damage regulated autophagy modulator 2
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_004624772: 6899536 - 6922918 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05100148_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dram2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_013075505.2 XP_012930959.1 8 - 9 980 87
XM_013075509.2 XP_012930963.1 8 - 9 878 87
XM_021265644.1 XP_021121303.1 9 - 10 1067 87
XM_021265645.1 XP_021121304.1 9 - 10 1006 87
XM_021265646.1 XP_021121305.1 9 - 10 1052 87
XM_021265647.1 XP_021121306.1 9 - 10 969 87
XM_021265648.1 XP_021121307.1 8 - 9 948 87
XM_021265649.1 XP_021121308.1 9 - 10 1064 87
XM_021265650.1 XP_021121309.1 10 - 11 1223 87
XM_021265651.1 XP_021121310.1 8 - 9 848 87
XM_021265652.1 XP_021121311.1 8 - 9 945 87
XM_021265653.1 XP_021121312.1 9 - 10 1104 87
XM_021265654.1 XP_021121313.1 8 - 9 845 87
XM_021265655.1 XP_021121314.1 9 - 10 1004 87
XM_021265656.1 XP_021121315.1 7 - 8 796 87

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0