This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05101842_g1
See other CAPG GE Assays ›
Gene Symbol:
Capg
Gene Name:
capping actin protein, gelsolin like
Gene Aliases:
GW7_02422
Chromosome Location:
Chr.Un|NW_004624749: 17706501 - 17746346 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05101842_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Capg
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_021258219.1 XP_021113878.1 8 - 9 977 59
XM_021258220.1 XP_021113879.1 9 - 10 1098 59
XM_021258221.1 XP_021113880.1 8 - 9 1088 59
XM_021258222.1 XP_021113881.1 9 - 10 1141 59
XM_021258223.1 XP_021113882.1 8 - 9 1028 59
XM_021258224.1 XP_021113883.1 8 - 9 933 59
XM_021258225.1 XP_021113884.1 8 - 9 1065 59
XM_021258226.1 XP_021113885.1 8 - 9 946 59
XM_021258227.1 XP_021113886.1 8 - 9 930 59
XM_021258228.1 XP_021113887.1 8 - 9 1096 59
XM_021258229.1 XP_021113888.1 8 - 9 932 59
XM_021258230.1 XP_021113889.1 7 - 8 908 59
XM_021258231.1 XP_021113890.1 7 - 8 908 59
XM_021258232.1 XP_021113891.1 7 - 8 908 59
XM_021258233.1 XP_021113892.1 7 - 8 908 59
XM_021258234.1 XP_021113893.1 5 - 6 665 59

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

non-motor actin binding protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-kt9qp:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0