This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05106148_m1
See other DTX2 GE Assays ›
Gene Symbol:
Dtx2
Gene Name:
deltex E3 ubiquitin ligase 2
Gene Aliases:
GW7_12436
Chromosome Location:
Chr.Un|NW_004624740: 15056018 - 15098322 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05106148_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dtx2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004840100.3 XP_004840157.1 7 - 8 1957 66
XM_004840105.3 XP_004840162.1 6 - 7 1527 66
XM_004840106.2 XP_004840163.1 6 - 7 1479 66
XM_021254440.1 XP_021110099.1 9 - 10 1972 66
XM_021254441.1 XP_021110100.1 9 - 10 1944 66
XM_021254442.1 XP_021110101.1 9 - 10 1941 66
XM_021254443.1 XP_021110102.1 6 - 7 1332 66
XM_021254444.1 XP_021110103.1 5 - 6 1222 66
XM_021254445.1 XP_021110104.1 4 - 5 1081 66

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jmd5k:80/100.66.78.41:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0