This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05110378_m1
See other AASDH GE Assays ›
Gene Symbol:
Aasdh
Gene Name:
aminoadipate-semialdehyde dehydrogenase
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_004624761: 13026881 - 13063202 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05110378_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Aasdh
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004849696.3 XP_004849753.2 15 - 16 3190 79
XM_013073948.2 XP_012929402.1 14 - 15 3020 79
XM_013073949.2 XP_012929403.1 10 - 11 2276 79
XM_013073951.2 XP_012929405.1 8 - 9 1850 79
XM_021262315.1 XP_021117974.1 15 - 16 3147 79
XM_021262316.1 XP_021117975.1 15 - 16 3282 79
XM_021262317.1 XP_021117976.1 15 - 16 3087 79
XM_021262318.1 XP_021117977.1 13 - 14 2838 79
XM_021262319.1 XP_021117978.1 15 - 16 2748 79

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

ligase metabolite interconversion enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0