This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05111914_m1
See other RSG1 GE Assays ›
Gene Symbol:
Rsg1
Gene Name:
REM2 and RAB like small GTPase 1
Gene Aliases:
GW7_15528
Chromosome Location:
Chr.Un|NW_004624764: 2350780 - 2358465 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05111914_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rsg1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004850363.2 XP_004850420.1 2 - 3 262 94
XM_013074365.2 XP_012929819.1 3 - 4 963 94
XM_021262868.1 XP_021118527.1 3 - 4 939 94

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-55658555d6-9rjdb:80/100.66.79.224:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline