This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05112198_gH
See other NAA10 GE Assays ›
Gene Symbol:
Naa10
Gene Name:
N(alpha)-acetyltransferase 10, NatA catalytic subunit
Gene Aliases:
Renbp
Chromosome Location:
Chr.Un|NW_004624946: 565221 - 570330 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05112198_gH
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Naa10
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004874972.3 XP_004875029.1 2 - 3 234 61
XM_021252306.1 XP_021107965.1 11 - 12 1378 61
XM_021252307.1 XP_021107966.1 11 - 12 1353 61
XM_021252308.1 XP_021107967.1 11 - 12 1344 61
XM_021252309.1 XP_021107968.1 11 - 12 1369 61
XM_021252310.1 XP_021107969.1 11 - 12 1353 61
XM_021252311.1 XP_021107970.1 10 - 11 1243 61
XM_021252313.1 XP_021107972.1 10 - 11 1128 61

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_004874972.3 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

acetyltransferase epimerase/racemase isomerase metabolite interconversion enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-kt9qp:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0