This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05114553_g1
See other ROMO1 GE Assays ›
Gene Symbol:
Romo1
Gene Name:
reactive oxygen species modulator 1
Gene Aliases:
GW7_21269
Chromosome Location:
Chr.Un|NW_004624842: 3734441 - 3736354 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05114553_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Romo1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004867865.2 XP_004867922.1 1 - 2 115 78

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_004867865.2 transcripts.

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-55ff9ddd88-lm858:80/100.66.77.133:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0