This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05118357_g1
See other SLC50A1 GE Assays ›
Gene Symbol:
Slc50a1
Gene Name:
solute carrier family 50 member 1
Gene Aliases:
GW7_05197
Chromosome Location:
Chr.Un|NW_004624885: 2379489 - 2381918 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05118357_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc50a1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004871297.2 XP_004871354.1 1 - 2 170 61
XM_004871299.2 XP_004871356.1 1 - 2 171 61
XM_021249433.1 XP_021105092.1 1 - 2 171 61
XM_021249435.1 XP_021105094.1 1 - 2 171 61
XM_021249437.1 XP_021105096.1 1 - 2 171 61
XM_021249438.1 XP_021105097.1 1 - 2 171 61
XM_021249439.1 XP_021105098.1 1 - 2 170 61

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-hk65g:80/100.66.77.69:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0