This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05119638_m1
See other ARHGAP24 GE Assays ›
Gene Symbol:
Arhgap24
Gene Name:
Rho GTPase activating protein 24
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_004624872: 1950076 - 2442522 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05119638_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Arhgap24
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004870505.3 XP_004870562.1 6 - 7 962 71
XM_004870507.3 XP_004870564.1 3 - 4 1685 71
XM_004870508.3 XP_004870565.1 4 - 5 734 71
XM_004870509.3 XP_004870566.1 4 - 5 688 71
XM_004870510.3 XP_004870567.1 4 - 5 483 71
XM_021248782.1 XP_021104441.1 4 - 5 480 71

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0