Assay Details
| Gene Symbol: Lsm5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004872209.3 | XP_004872266.1 | 2 - 3 | 207 | 81 | |
| XM_021250162.1 | XP_021105821.1 | 3 - 4 | 436 | 81 | ||
| XM_021250163.1 | XP_021105822.1 | 3 - 4 | 296 | 81 | ||
More Information
Set Membership: |
|

Lsm5
Genomic Map