This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05121602_m1
See other KMT5B GE Assays ›
Gene Symbol:
Kmt5b
Gene Name:
lysine methyltransferase 5B
Gene Aliases:
GW7_09656, SUV420H1
Chromosome Location:
Chr.Un|NW_004624767: 18655192 - 18714254 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05121602_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Kmt5b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004852192.3 XP_004852249.1 7 - 8 1627 74
XM_004852194.3 XP_004852251.1 7 - 8 1086 74
XM_013074821.2 XP_012930275.1 8 - 9 1779 74
XM_013074822.2 XP_012930276.1 8 - 9 1768 74
XM_021264483.1 XP_021120142.1 7 - 8 1518 74
XM_021264484.1 XP_021120143.1 5 - 6 691 74

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0