This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05121872_m1
See other GMNN GE Assays ›
Gene Symbol:
Gmnn
Gene Name:
geminin, DNA replication inhibitor
Gene Aliases:
geminin
Chromosome Location:
Chr.Un|NW_004624756: 2268304 - 2335026 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05121872_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gmnn
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004847593.2 XP_004847650.1 5 - 6 655 74
XM_004847594.2 XP_004847651.1 5 - 6 652 74
XM_004847595.2 XP_004847652.1 6 - 7 747 74
XM_004847596.2 XP_004847653.1 3 - 4 572 74
XM_013073277.2 XP_012928731.1 3 - 4 569 74
XM_013073278.2 XP_012928732.1 3 - 4 318 74
XM_021260555.1 XP_021116214.1 6 - 7 744 74
XM_021260556.1 XP_021116215.1 4 - 5 585 74
XM_021260557.1 XP_021116216.1 3 - 4 313 74
XM_021260558.1 XP_021116217.1 5 - 6 660 74

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0