This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05123930_m1
See other RARA GE Assays ›
Gene Symbol:
Rara
Gene Name:
retinoic acid receptor alpha
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_004624795: 2934995 - 2975236 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05123930_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rara
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004859443.3 XP_004859500.1 3 - 4 1186 58
XM_004859447.3 XP_004859504.1 4 - 5 972 58
XM_004859448.3 XP_004859505.1 3 - 4 796 58
XM_004859449.3 XP_004859506.2 3 - 4 798 58
XM_013077738.2 XP_012933192.1 4 - 5 900 58
XM_013077739.2 XP_012933193.1 3 - 4 396 58
XM_021239972.1 XP_021095631.1 3 - 4 308 58

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

C4 zinc finger nuclear receptor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0