This product has been added to your favorites list. Go to
My Favorites
Please enter the information below and press OK to send your cart to Core Services for purchase.
Name
Phone number
Email
Budget number
PI Name
PI Email
Building and Room number
Pickup
Delivery
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Hg05124427_m1
See other ATG4C GE Assays ›
Gene Symbol:
Atg4c
Gene Name:
autophagy related 4C cysteine peptidase
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_004624742: 31061869 - 31131179 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID
Hg05124427_m1
Best Coverage
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: Atg4c
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
XM_004841539.3
XP_004841596.1
6 - 7
989
63
XM_013071007.2
XP_012926461.1
9 - 10
1443
63
XM_013071008.2
XP_012926462.1
9 - 10
1411
63
XM_013071009.2
XP_012926463.1
7 - 8
6592
63
XM_013071010.2
XP_012926464.1
9 - 10
1585
63
XM_013071013.2
XP_012926467.1
8 - 9
1422
63
XM_013071014.2
XP_012926468.1
8 - 9
1735
63
XM_013071015.2
XP_012926469.1
9 - 10
1367
63
XM_013071016.2
XP_012926470.1
8 - 9
1730
63
XM_013071017.2
XP_012926471.1
8 - 9
1395
63
XM_013071018.2
XP_012926472.1
8 - 9
1244
63
XM_013071020.2
XP_012926474.1
7 - 8
1168
63
XM_013071021.2
XP_012926475.1
5 - 6
939
63
XM_013071024.2
XP_012926478.1
9 - 10
1467
63
XM_021255624.1
XP_021111283.1
8 - 9
1573
63
XM_021255625.1
XP_021111284.1
8 - 9
1364
63
XM_021255626.1
XP_021111285.1
8 - 9
1288
63
XM_021255627.1
XP_021111286.1
8 - 9
1343
63
XM_021255628.1
XP_021111287.1
7 - 8
1263
63
XM_021255629.1
XP_021111288.1
6 - 7
1064
63
XM_021255630.1
XP_021111289.1
4 - 5
841
63
XM_021255631.1
XP_021111290.1
9 - 10
1459
63
Back To Top
More Information
Set Membership:
Amplicon length less than or equal to 70
Recommended Assay
Probe spans exons
Panther Classification:
Molecular Function -
cysteine protease
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0