This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05124615_m1
See other METTL15 GE Assays ›
Gene Symbol:
Mettl15
Gene Name:
methyltransferase like 15
Gene Aliases:
GW7_11697
Chromosome Location:
Chr.Un|NW_004624766: 17624565 - 17786813 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05124615_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mettl15
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004851585.3 XP_004851642.1 3 - 4 588 63
XM_004851586.3 XP_004851643.2 3 - 4 522 63
XM_021263955.1 XP_021119614.1 3 - 4 522 63
XM_021263956.1 XP_021119615.1 3 - 4 522 63
XM_021263957.1 XP_021119616.1 3 - 4 522 63
XM_021263958.1 XP_021119617.1 3 - 4 522 63
XM_021263959.1 XP_021119618.1 3 - 4 522 63

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

methyltransferase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-wt24k:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline