This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05126371_g1
See other YIF1B GE Assays ›
Gene Symbol:
Yif1b
Gene Name:
Yip1 interacting factor homolog B, membrane trafficking protein
Gene Aliases:
GW7_16722
Chromosome Location:
Chr.Un|NW_004624794: 11774494 - 11783383 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05126371_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Yif1b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004859039.3 XP_004859096.1 7 - 8 964 60
XM_004859040.3 XP_004859097.1 7 - 8 869 60
XM_004859041.3 XP_004859098.1 7 - 8 1305 60
XM_004859042.3 XP_004859099.1 8 - 9 1091 60
XM_021239835.1 XP_021095494.1 8 - 9 1224 60
XM_021239836.1 XP_021095495.1 7 - 8 974 60
XM_021239837.1 XP_021095496.1 8 - 9 1070 60

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-vh84l:80/100.66.79.51:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0