This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05126819_m1
See other EPHX1 GE Assays ›
Gene Symbol:
Ephx1
Gene Name:
epoxide hydrolase 1
Gene Aliases:
GW7_03770
Chromosome Location:
Chr.Un|NW_004624835: 6161930 - 6180981 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05126819_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ephx1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_013066506.2 XP_012921960.1 10 - 11 1969 58
XM_013066508.2 XP_012921962.1 8 - 9 1798 58
XM_013066510.2 XP_012921964.1 8 - 9 1311 58
XM_013066511.2 XP_012921965.1 8 - 9 1299 58
XM_013066512.2 XP_012921966.1 8 - 9 1503 58
XM_021246475.1 XP_021102134.1 9 - 10 1508 58

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

hydrolase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline