This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05128381_m1
See other NDUFAF7 GE Assays ›
Gene Symbol:
Ndufaf7
Gene Name:
NADH:ubiquinone oxidoreductase complex assembly factor 7
Gene Aliases:
GW7_21645
Chromosome Location:
Chr.Un|NW_004624738: 18836543 - 18854474 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05128381_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ndufaf7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004839301.2 XP_004839358.1 8 - 9 1014 82
XM_004839302.2 XP_004839359.1 7 - 8 801 82
XM_021253730.1 XP_021109389.1 9 - 10 1176 82
XM_021253731.1 XP_021109390.1 9 - 10 1217 82
XM_021253732.1 XP_021109391.1 9 - 10 966 82
XM_021253733.1 XP_021109392.1 8 - 9 963 82
XM_021253734.1 XP_021109393.1 7 - 8 903 82
XM_021253735.1 XP_021109394.1 8 - 9 1049 82

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-fcf5f9d6d-l2zpb:80/100.66.75.138:80.
git-commit: 856de8fa7eff2d55603d92afdf035972e45c3e8a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.51.0-2026.05.21-1.0