This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05129840_m1
See other HADHB GE Assays ›
Gene Symbol:
Hadhb
Gene Name:
hydroxyacyl-CoA dehydrogenase trifunctional multienzyme complex subunit beta
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_004624738: 8384819 - 8420743 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05129840_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Hadhb
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004839132.2 XP_004839189.1 11 - 12 1328 62
XM_004839133.2 XP_004839190.1 10 - 11 1216 62
XM_004839134.2 XP_004839191.1 10 - 11 1052 62
XM_004839135.2 XP_004839192.1 10 - 11 1098 62
XM_004839136.2 XP_004839193.1 11 - 12 1128 62
XM_021253587.1 XP_021109246.1 9 - 10 996 62

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

acyltransferase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0