This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05130464_g1
See other FAM173B GE Assays ›
Gene Symbol:
Fam173b
Gene Name:
family with sequence similarity 173 member B
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_004624751: 7598313 - 7614801 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05130464_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Fam173b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004845178.2 XP_004845235.2 1 - 2 182 101
XM_013072376.2 XP_012927830.2 1 - 2 182 101
XM_013072378.2 XP_012927832.2 1 - 2 182 101
XM_013072379.2 XP_012927833.2 1 - 2 182 101
XM_021258612.1 XP_021114271.1 1 - 2 182 101
XM_021258613.1 XP_021114272.1 1 - 2 182 101
XM_021258614.1 XP_021114273.1 1 - 2 182 101
XM_021258615.1 XP_021114274.1 1 - 2 182 101
XM_021258616.1 XP_021114275.1 1 - 2 182 101

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline