This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05131692_m1
See other PDGFA GE Assays ›
Gene Symbol:
Pdgfa
Gene Name:
platelet derived growth factor subunit A
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_004624740: 26172868 - 26189629 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05131692_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pdgfa
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004840314.3 XP_004840371.2 3 - 4 499 74
XM_013070566.2 XP_012926020.1 3 - 4 454 74
XM_021254603.1 XP_021110262.1 4 - 5 570 74
XM_021254604.1 XP_021110263.1 4 - 5 568 74
XM_021254605.1 XP_021110264.1 4 - 5 657 74
XM_021254606.1 XP_021110265.1 3 - 4 457 74
XM_021254607.1 XP_021110266.1 4 - 5 354 74
XM_021254608.1 XP_021110267.1 4 - 5 444 74

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

growth factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline