This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05131870_g1
See other PPIB GE Assays ›
Gene Symbol:
Ppib
Gene Name:
peptidylprolyl isomerase B
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_004624781: 8402545 - 8407947 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05131870_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ppib
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004855661.3 XP_004855718.1 3 - 4 407 61

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Endogenous Controls Detect Genomic DNA Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

chaperone

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline