This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05134501_m1
See other NDUFA11 GE Assays ›
Gene Symbol:
Ndufa11
Gene Name:
NADH:ubiquinone oxidoreductase subunit A11
Gene Aliases:
GW7_20484
Chromosome Location:
Chr.Un|NW_004624828: 4319665 - 4328267 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05134501_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ndufa11
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004865660.3 XP_004865717.1 2 - 3 294 59
XM_004865661.2 XP_004865718.1 2 - 3 294 59

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

oxidoreductase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-fcf5f9d6d-dcxfx:80/100.66.77.77:80.
git-commit: 856de8fa7eff2d55603d92afdf035972e45c3e8a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.51.0-2026.05.21-1.0