Assay Details
| Gene Symbol: Smarcb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004843579.3 | XP_004843636.1 | 4 - 5 | 702 | 59 | |
| XM_004843580.3 | XP_004843637.1 | 4 - 5 | 674 | 59 | ||
More Information
Set Membership: |
|

Smarcb1
Genomic Map