Assay Details
| Gene Symbol: Nqo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004842935.3 | XP_004842992.1 | 3 - 4 | 477 | 59 | |
| XM_004842936.3 | XP_004842993.1 | 3 - 4 | 477 | 59 | ||
| XM_021256778.1 | XP_021112437.1 | 4 - 5 | 408 | 59 | ||
| XM_021256779.1 | XP_021112438.1 | 3 - 4 | 392 | 59 | ||
More Information
Set Membership: |
|

Nqo1
Genomic Map