Assay Details
Gene Symbol: Tp53i11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004851980.3 | XP_004852037.1 | 7 - 8 | 692 | 63 | |
XM_004851981.3 | XP_004852038.1 | 7 - 8 | 1041 | 63 | ||
XM_013074860.2 | XP_012930314.1 | 7 - 8 | 697 | 63 | ||
XM_013074861.2 | XP_012930315.1 | 7 - 8 | 1049 | 63 | ||
XM_021264314.1 | XP_021119973.1 | 7 - 8 | 743 | 63 | ||
XM_021264315.1 | XP_021119974.1 | 7 - 8 | 652 | 63 | ||
XM_021264316.1 | XP_021119975.1 | 7 - 8 | 696 | 63 | ||
XM_021264317.1 | XP_021119976.1 | 7 - 8 | 608 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
