Assay Details
| Gene Symbol: Srd5a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004845156.3 | XP_004845213.1 | 1 - 2 | 457 | 67 | |
| XM_004845157.3 | XP_004845214.1 | 1 - 2 | 446 | 67 | ||
| XM_021258595.1 | XP_021114254.1 | 1 - 2 | 484 | 67 | ||
| XM_021258596.1 | XP_021114255.1 | 1 - 2 | 476 | 67 | ||
More Information
Set Membership: |
|

Srd5a1
Genomic Map