This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05141357_m1
See other MAMSTR GE Assays ›
Gene Symbol:
Mamstr
Gene Name:
MEF2 activating motif and SAP domain containing transcriptional regulator
Gene Aliases:
GW7_13636
Chromosome Location:
Chr.Un|NW_004624832: 5190976 - 5196665 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05141357_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mamstr
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004866985.2 XP_004867042.1 2 - 3 582 69
XM_021246190.1 XP_021101849.1 2 - 3 599 69
XM_021246192.1 XP_021101851.1 2 - 3 582 69
XM_021246193.1 XP_021101852.1 2 - 3 597 69
XM_021246194.1 XP_021101853.1 2 - 3 582 69
XM_021246195.1 XP_021101854.1 2 - 3 608 69
XM_021246196.1 XP_021101855.1 2 - 3 621 69
XM_021246197.1 XP_021101856.1 2 - 3 582 69

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0