This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hg05141376_g1
See other DYDC1 GE Assays ›
Gene Symbol:
Dydc1
Gene Name:
DPY30 domain containing 1
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_004624841: 6805847 - 6827463 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID Hg05141376_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dydc1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004867752.2 XP_004867809.1 6 - 7 691 81
XM_013066652.2 XP_012922106.1 7 - 8 692 81
XM_013066654.2 XP_012922108.1 6 - 7 608 81
XM_021246760.1 XP_021102419.1 7 - 8 703 81
XM_021246761.1 XP_021102420.1 6 - 7 609 81
XM_021246762.1 XP_021102421.1 6 - 7 1247 81
XM_021246763.1 XP_021102422.1 6 - 7 688 81
XM_021246764.1 XP_021102423.1 5 - 6 598 81

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-97tfg:80/100.66.76.242:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline